Assay Details
| Gene Symbol: wnt5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001362273.1 | NP_001349202.1 | 6 - 7 | 1548 | 70 | |
| NM_130937.2 | NP_571012.1 | 6 - 7 | 1553 | 70 | ||
| XM_017354945.2 | XP_017210434.1 | 3 - 4 | 1031 | 70 | ||
| GenBank mRNA | BC162981.1 | - | 3 - 4 | 684 | 70 | |
| BC162984.1 | - | 3 - 4 | 684 | 70 | ||
| U51268.1 | - | 5 - 6 | 824 | 70 | ||
More Information
Set Membership: |
|

wnt5b
Genomic Map