Assay Details
| Gene Symbol: pou4f3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131278.1 | NP_571353.1 | 1 - 2 | 97 | 61 | |
| GenBank mRNA | AY995217.1 | - | 1 - 2 | 97 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pou4f3| Gene Symbol: pou4f3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131278.1 | NP_571353.1 | 1 - 2 | 97 | 61 | |
| GenBank mRNA | AY995217.1 | - | 1 - 2 | 97 | 61 | |
Set Membership: |
|