Assay Details
| Gene Symbol: has2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153650.2 | NP_705936.1 | 3 - 4 | 1261 | 67 | |
| GenBank mRNA | AF190742.1 | - | 3 - 4 | 761 | 67 | |
| BC100130.1 | - | 3 - 4 | 1261 | 67 | ||
| U53223.1 | - | 1 - 2 | 86 | 67 | ||
More Information
Set Membership: |
|

has2
Genomic Map