Assay Details
| Gene Symbol: thbs3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173225.1 | NP_775332.1 | 16 - 17 | 1970 | 135 | |
| GenBank mRNA | AF462310.1 | - | 16 - 17 | 1970 | 135 | |
| BC085524.1 | - | 16 - 17 | 1968 | 135 | ||
| BC165677.1 | - | 16 - 17 | 1920 | 135 | ||
More Information
Set Membership: |
|

thbs3a
Genomic Map