Assay Details
| Gene Symbol: thbs3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173225.1 | NP_775332.1 | 8 - 9 | 1054 | 68 | |
| GenBank mRNA | AF462310.1 | - | 8 - 9 | 1054 | 68 | |
More Information
Set Membership: |
|

thbs3a
Genomic Map