Assay Details
| Gene Symbol: mc1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180970.1 | NP_851301.1 | 1 | 847 | 96 | |
| XM_005159179.4 | XP_005159236.1 | 2 | 1108 | 96 | ||
| GenBank mRNA | BC162836.1 | - | 1 | 881 | 96 | |
| BC162848.1 | - | 1 | 881 | 96 | ||
More Information
Set Membership: |
|

mc1r
Genomic Map