Assay Details
| Gene Symbol: rpl5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199756.2 | NP_956050.1 | 3 - 4 | 268 | 83 | |
| GenBank mRNA | BC049035.1 | - | 3 - 4 | 272 | 83 | |
| BC065687.1 | - | 3 - 4 | 251 | 83 | ||
More Information
Set Membership: |
|

rpl5a
Genomic Map