Assay Details
| Gene Symbol: rnps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199761.2 | 6 - 7 | 972 | 71 | ||
| XM_009298275.3 | XP_009296550.1 | 6 - 7 | 938 | 71 | ||
| GenBank mRNA | BC057251.1 | - | 6 - 7 | 798 | 71 | |
| BC065953.1 | - | 6 - 7 | 783 | 71 | ||
More Information
Set Membership: |
|

rnps1
Genomic Map