Assay Details
| Gene Symbol: dcun1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199772.1 | NP_956066.1 | 6 - 7 | 749 | 91 | |
| GenBank mRNA | BC057530.1 | - | 6 - 7 | 749 | 91 | |
| BC068381.1 | - | 6 - 7 | 715 | 91 | ||
More Information
Set Membership: |
|

dcun1d1
Genomic Map