Assay Details
| Gene Symbol: trim25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200175.1 | NP_956469.1 | 9 - 10 | 1433 | 95 | |
| XM_017356449.2 | XP_017211938.1 | 8 - 9 | 1427 | 95 | ||
| GenBank mRNA | BC045450.1 | - | 9 - 10 | 1433 | 95 | |
| BC165638.1 | - | 9 - 10 | 1416 | 95 | ||
More Information
Set Membership: |
|

trim25
Genomic Map