Assay Details
| Gene Symbol: prelid1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200366.2 | NP_956660.1 | 2 - 3 | 242 | 93 | |
| GenBank mRNA | AY398403.1 | - | 2 - 3 | 242 | 93 | |
| BC053211.1 | - | 2 - 3 | 192 | 93 | ||
| BC165914.1 | - | 1 - 2 | 124 | 93 | ||
More Information
Set Membership: |
|

prelid1a
Genomic Map