Assay Details
| Gene Symbol: prelid1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200366.2 | NP_956660.1 | 4 - 5 | 583 | 97 | |
| GenBank mRNA | AY398403.1 | - | 4 - 5 | 583 | 97 | |
| BC053211.1 | - | 4 - 5 | 533 | 97 | ||
| BC165914.1 | - | 3 - 4 | 465 | 97 | ||
More Information
Set Membership: |
|

prelid1a
Genomic Map