Assay Details
| Gene Symbol: ppan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201463.1 | NP_958871.1 | 7 - 8 | 826 | 65 | |
| GenBank mRNA | BC045506.1 | - | 7 - 8 | 826 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ppan| Gene Symbol: ppan | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201463.1 | NP_958871.1 | 7 - 8 | 826 | 65 | |
| GenBank mRNA | BC045506.1 | - | 7 - 8 | 826 | 65 | |
Set Membership: |
|