Assay Details
| Gene Symbol: nrp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205674.1 | NP_991237.1 | 1543 | 78 | ||
| GenBank mRNA | AY351911.1 | - | 1557 | 78 | ||
| AY428799.1 | - | 1543 | 78 | |||
| AY428800.1 | - | 1543 | 78 | |||
| AY493415.1 | - | 1356 | 78 | |||
| BC133730.1 | - | 1521 | 78 | |||
| KM655608.1 | - | 1136 | 78 | |||
More Information
Set Membership: |
|

nrp1b
Genomic Map