Assay Details
| Gene Symbol: nrp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205674.1 | NP_991237.1 | 2880 | 70 | ||
| GenBank mRNA | AY351911.1 | - | 2894 | 70 | ||
| AY428799.1 | - | 2880 | 70 | |||
| AY428800.1 | - | 2880 | 70 | |||
| AY493415.1 | - | 2693 | 70 | |||
| KM655608.1 | - | 2473 | 70 | |||
| Gene Symbol: LOC110438108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021467877.1 | XP_021323552.1 | 2 - 3 | 514 | 70 | |
More Information
Set Membership: |
|

nrp1b
Genomic Map