Assay Details
| Gene Symbol: nrp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205674.1 | NP_991237.1 | 2971 | 117 | ||
| GenBank mRNA | AY351911.1 | - | 2985 | 117 | ||
| AY428799.1 | - | 2971 | 117 | |||
| AY493415.1 | - | 2784 | 117 | |||
| BC133730.1 | - | 2949 | 117 | |||
| KM655608.1 | - | 2564 | 117 | |||
| Gene Symbol: LOC110438108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021467877.1 | XP_021323552.1 | 3 - 4 | 605 | 117 | |
More Information
Set Membership: |
|

nrp1b
Genomic Map