Assay Details
| Gene Symbol: sypl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212669.1 | NP_997834.1 | 5 - 6 | 648 | 72 | |
| XM_017357386.2 | XP_017212875.1 | 4 - 5 | 606 | 72 | ||
| GenBank mRNA | AY394944.1 | - | 5 - 6 | 648 | 72 | |
| BC092895.1 | - | 5 - 6 | 653 | 72 | ||
| BC154773.1 | - | 5 - 6 | 643 | 72 | ||
More Information
Set Membership: |
|

sypl2b
Genomic Map