Assay Details
| Gene Symbol: ugp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316890.1 | NP_001303819.1 | 5 - 6 | 916 | 77 | |
| NM_212729.2 | NP_997894.2 | 5 - 6 | 604 | 77 | ||
| XM_005158496.4 | XP_005158553.1 | 5 - 6 | 964 | 77 | ||
| GenBank mRNA | BC054939.1 | - | 5 - 6 | 622 | 77 | |
| BC067564.1 | - | 5 - 6 | 604 | 77 | ||
| EH433688.1 | - | 5 - 6 | 643 | 77 | ||
More Information
Set Membership: |
|

ugp2b
Genomic Map