Assay Details
| Gene Symbol: lactb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212884.1 | NP_998049.1 | 374 | 64 | ||
| GenBank mRNA | BC066620.1 | - | 374 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

lactb2| Gene Symbol: lactb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212884.1 | NP_998049.1 | 374 | 64 | ||
| GenBank mRNA | BC066620.1 | - | 374 | 64 | ||
Set Membership: |
|