Assay Details
| Gene Symbol: rpl19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213208.1 | NP_998373.1 | 3 - 4 | 259 | 141 | |
| GenBank mRNA | AY648758.1 | - | 3 - 4 | 265 | 141 | |
| BC062844.1 | - | 3 - 4 | 259 | 141 | ||
More Information
Set Membership: |
|

rpl19
Genomic Map