Assay Details
| Gene Symbol: asic1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214791.1 | NP_999956.1 | 4 - 5 | 1185 | 77 | |
| XM_005161666.4 | XP_005161723.1 | 4 - 5 | 1469 | 77 | ||
| XM_005161667.4 | XP_005161724.1 | 5 - 6 | 1921 | 77 | ||
| GenBank mRNA | AJ609615.1 | - | 4 - 5 | 1185 | 77 | |
| BC162362.1 | - | 4 - 5 | 1154 | 77 | ||
| BC162368.1 | - | 4 - 5 | 1154 | 77 | ||
More Information
Set Membership: |
|

asic1b
Genomic Map