Assay Details
| Gene Symbol: rbmx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025166.1 | NP_001020337.1 | 6 | 651 | 90 | |
| XM_005170268.4 | XP_005170325.1 | 6 | 90 | |||
| GenBank mRNA | BC078245.1 | - | 6 | 694 | 90 | |
| BC096985.1 | - | 6 | 651 | 90 | ||
More Information
Set Membership: |
|

rbmx2
Genomic Map