Assay Details
| Gene Symbol: stk17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001089337.1 | NP_001082806.1 | 7 | 3152 | 90 | |
| GenBank mRNA | BC134810.1 | - | 7 | 3152 | 90 | |
More Information
Set Membership: |
|

stk17a
Genomic Map