Assay Details
| Gene Symbol: eif4ebp3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199683.1 | NP_955977.1 | 3 | 1056 | 154 | |
| GenBank mRNA | BC046079.1 | - | 3 | 1056 | 154 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eif4ebp3l| Gene Symbol: eif4ebp3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199683.1 | NP_955977.1 | 3 | 1056 | 154 | |
| GenBank mRNA | BC046079.1 | - | 3 | 1056 | 154 | |
Set Membership: |
|