Assay Details
| Gene Symbol: scfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013564.2 | NP_001013582.1 | 6 | 1744 | 86 | |
| GenBank mRNA | BC090478.1 | - | 6 | 1748 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

scfd2| Gene Symbol: scfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013564.2 | NP_001013582.1 | 6 | 1744 | 86 | |
| GenBank mRNA | BC090478.1 | - | 6 | 1748 | 86 | |
Set Membership: |
|