Assay Details
| Gene Symbol: mpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082850.1 | NP_001076319.1 | 1 | 445 | 139 | |
| GenBank mRNA | BC134182.1 | - | 1 | 445 | 139 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mpg| Gene Symbol: mpg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082850.1 | NP_001076319.1 | 1 | 445 | 139 | |
| GenBank mRNA | BC134182.1 | - | 1 | 445 | 139 | |
Set Membership: |
|