Assay Details
| Gene Symbol: btg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130922.2 | NP_570997.1 | 2 | 654 | 147 | |
| GenBank mRNA | AB036784.1 | - | 2 | 649 | 147 | |
| BC044343.1 | - | 2 | 647 | 147 | ||
| BC097098.1 | - | 2 | 647 | 147 | ||
| BC163955.1 | - | 2 | 495 | 147 | ||
| EH434869.1 | - | 2 | 660 | 147 | ||
More Information
Set Membership: |
|

btg2
Genomic Map