Assay Details
| Gene Symbol: cfl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213641.2 | NP_998806.1 | 2 | 190 | 130 | |
| GenBank mRNA | AY398324.1 | - | 2 | 152 | 130 | |
| BC067336.1 | - | 2 | 120 | 130 | ||
| EH547447.1 | - | 2 | 181 | 130 | ||
More Information
Set Membership: |
|

cfl1
Genomic Map