Assay Details
| Gene Symbol: mfng | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007788.1 | NP_001007789.1 | 8 | 1752 | 102 | |
| GenBank mRNA | AY608926.1 | - | 8 | 1752 | 102 | |
| BC125890.1 | - | 8 | 1737 | 102 | ||
More Information
Set Membership: |
|

mfng
Genomic Map