Assay Details
| Gene Symbol: prim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214782.2 | NP_999947.1 | 13 | 1850 | 99 | |
| XM_005156811.4 | XP_005156868.1 | 14 | 1557 | 99 | ||
| GenBank mRNA | AY398398.1 | - | 14 | 1528 | 99 | |
| BC095387.1 | - | 13 | 1850 | 99 | ||
More Information
Set Membership: |
|

prim2
Genomic Map