Assay Details
| Gene Symbol: rarga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131339.1 | NP_571414.1 | 8 | 1655 | 133 | |
| GenBank mRNA | BC098519.1 | - | 8 | 1663 | 133 | |
| S74156.1 | - | 8 | 1655 | 133 | ||
More Information
Set Membership: |
|

rarga
Genomic Map