Assay Details
| Gene Symbol: ncam1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131202.2 | NP_571277.2 | 10 | 1461 | 119 | |
| XM_005161135.4 | XP_005161192.1 | 11 | 1521 | 119 | ||
| XM_005161137.4 | XP_005161194.1 | 11 | 1521 | 119 | ||
| XM_005161138.4 | XP_005161195.1 | 11 | 1521 | 119 | ||
| XM_005161139.4 | XP_005161196.1 | 10 | 1490 | 119 | ||
| XM_005161141.4 | XP_005161198.2 | 11 | 1521 | 119 | ||
| XM_005161142.4 | XP_005161199.1 | 10 | 1490 | 119 | ||
| XM_009295280.3 | XP_009293555.1 | 11 | 1522 | 119 | ||
| XM_009295281.3 | XP_009293556.1 | 10 | 1491 | 119 | ||
| XM_009295282.3 | XP_009293557.1 | 11 | 1521 | 119 | ||
| XM_009295286.3 | XP_009293561.1 | 11 | 1521 | 119 | ||
| XM_009295288.3 | XP_009293563.1 | 11 | 1523 | 119 | ||
| XM_017352744.2 | XP_017208233.1 | 10 | 1491 | 119 | ||
| XM_017352745.2 | XP_017208234.1 | 11 | 1521 | 119 | ||
| XM_017352746.2 | XP_017208235.1 | 10 | 1490 | 119 | ||
| GenBank mRNA | AY029402.1 | - | 10 | 1450 | 119 | |
| BC162675.1 | - | 10 | 1421 | 119 | ||
| CU458885.1 | - | 10 | 1320 | 119 | ||
| HM467818.1 | - | 10 | 1320 | 119 | ||
More Information
Set Membership: |
|

ncam1a
Genomic Map