Assay Details
| Gene Symbol: neurl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083831.1 | NP_001077300.1 | 2 | 1541 | 135 | |
| GenBank mRNA | BC134917.1 | - | 2 | 1541 | 135 | |
More Information
Set Membership: |
|

neurl2
Genomic Map