Assay Details
| Gene Symbol: isg20l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103193.2 | NP_001096663.1 | 4 | 1245 | 143 | |
| XM_009292339.3 | XP_009290614.1 | 3 | 576 | 143 | ||
| GenBank mRNA | BC152518.1 | - | 4 | 1248 | 143 | |
More Information
Set Membership: |
|

isg20l2
Genomic Map