Assay Details
| Gene Symbol: arf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199275.1 | NP_954969.1 | 6 | 1655 | 147 | |
| GenBank mRNA | BC047804.1 | - | 6 | 1655 | 147 | |
| BC153642.1 | - | 6 | 1606 | 147 | ||
More Information
Set Membership: |
|

arf5
Genomic Map