Assay Details
| Gene Symbol: capn5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080007.1 | NP_001073476.1 | 10 | 1854 | 135 | |
| GenBank mRNA | BC125867.1 | - | 10 | 1854 | 135 | |
| BC164555.1 | - | 9 | 1437 | 135 | ||
More Information
Set Membership: |
|

capn5a
Genomic Map