Assay Details
| Gene Symbol: serpina7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105589.3 | NP_001099059.2 | 3 | 803 | 121 | |
| XM_021476265.1 | XP_021331940.1 | 2 | 805 | 121 | ||
| GenBank mRNA | BC153540.1 | - | 2 | 754 | 121 | |
More Information
Set Membership: |
|

serpina7
Genomic Map