Assay Details
| Gene Symbol: mansc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083027.2 | NP_001076496.1 | 4 | 845 | 128 | |
| XM_005168490.4 | XP_005168547.1 | 4 | 894 | 128 | ||
| XM_005168491.4 | XP_005168548.1 | 4 | 765 | 128 | ||
| XM_005168492.4 | XP_005168549.1 | 4 | 840 | 128 | ||
| GenBank mRNA | BC133089.1 | - | 4 | 849 | 128 | |
More Information
Set Membership: |
|

mansc1
Genomic Map