Assay Details
| Gene Symbol: pnpla7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080655.1 | NP_001074124.1 | 10 | 1049 | 130 | |
| GenBank mRNA | BC129427.1 | - | 10 | 1049 | 130 | |
| BC164358.1 | - | 9 | 887 | 130 | ||
More Information
Set Membership: |
|

pnpla7a
Genomic Map