Assay Details
| Gene Symbol: klhl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083022.1 | NP_001076491.1 | 12 | 2945 | 113 | |
| XM_021466572.1 | XP_021322247.1 | 12 | 3023 | 113 | ||
| GenBank mRNA | BC133175.1 | - | 12 | 2945 | 113 | |
More Information
Set Membership: |
|

klhl32
Genomic Map