Assay Details
| Gene Symbol: ndufa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003516.3 | NP_001003516.2 | 4 | 774 | 93 | |
| GenBank mRNA | BC078251.1 | - | 4 | 751 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ndufa3| Gene Symbol: ndufa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003516.3 | NP_001003516.2 | 4 | 774 | 93 | |
| GenBank mRNA | BC078251.1 | - | 4 | 751 | 93 | |
Set Membership: |
|