Assay Details
| Gene Symbol: pawr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006015.2 | NP_001006015.1 | 6 | 1151 | 72 | |
| GenBank mRNA | BC083409.1 | - | 6 | 1151 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pawr| Gene Symbol: pawr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006015.2 | NP_001006015.1 | 6 | 1151 | 72 | |
| GenBank mRNA | BC083409.1 | - | 6 | 1151 | 72 | |
Set Membership: |
|