Assay Details
| Gene Symbol: agbl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017754.1 | NP_001017754.1 | 5 | 751 | 68 | |
| GenBank mRNA | BC093361.1 | - | 5 | 751 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

agbl4| Gene Symbol: agbl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017754.1 | NP_001017754.1 | 5 | 751 | 68 | |
| GenBank mRNA | BC093361.1 | - | 5 | 751 | 68 | |
Set Membership: |
|