Assay Details
| Gene Symbol: ptger4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039629.1 | NP_001034718.1 | 1 | 750 | 62 | |
| GenBank mRNA | BC127559.1 | - | 1 | 737 | 62 | |
| DQ202321.1 | - | 1 | 750 | 62 | ||
More Information
Set Membership: |
|

ptger4a
Genomic Map