Assay Details
| Gene Symbol: rnf126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083017.1 | NP_001076486.1 | 9 | 1549 | 134 | |
| GenBank mRNA | BC133164.1 | - | 9 | 1549 | 134 | |
More Information
Set Membership: |
|

rnf126
Genomic Map