Assay Details
| Gene Symbol: mafba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131015.3 | NP_571090.2 | 1 | 3255 | 106 | |
| GenBank mRNA | BC117640.1 | - | 1 | 2953 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mafba| Gene Symbol: mafba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131015.3 | NP_571090.2 | 1 | 3255 | 106 | |
| GenBank mRNA | BC117640.1 | - | 1 | 2953 | 106 | |
Set Membership: |
|