Assay Details
| Gene Symbol: snx12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145892.1 | NP_001139364.1 | 4 | 1731 | 148 | |
| NM_201123.2 | NP_957417.1 | 4 | 1758 | 148 | ||
| GenBank mRNA | BC044462.1 | - | 4 | 1746 | 148 | |
| BC124154.1 | - | 4 | 1713 | 148 | ||
| BC133125.1 | - | 4 | 1719 | 148 | ||
More Information
Set Membership: |
|

snx12
Genomic Map