Assay Details
| Gene Symbol: rab11bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002555.1 | NP_001002555.1 | 1 - 2 | 249 | 90 | |
| GenBank mRNA | BC076247.1 | - | 1 - 2 | 249 | 90 | |
| BC152630.1 | - | 1 - 2 | 213 | 90 | ||
More Information
Set Membership: |
|

rab11bb
Genomic Map