Assay Details
| Gene Symbol: dffa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002631.1 | NP_001002631.1 | 5 - 6 | 882 | 70 | |
| GenBank mRNA | BC075948.1 | - | 5 - 6 | 882 | 70 | |
More Information
Set Membership: |
|

dffa
Genomic Map