Assay Details
| Gene Symbol: cox6b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002695.3 | NP_001002695.1 | 3 - 4 | 382 | 115 | |
| GenBank mRNA | BC076119.1 | - | 3 - 4 | 308 | 115 | |
| EH487817.1 | - | 3 - 4 | 378 | 115 | ||
| EH500377.1 | - | 3 - 4 | 395 | 115 | ||
| EH583430.1 | - | 3 - 4 | 408 | 115 | ||
More Information
Set Membership: |
|

cox6b1
Genomic Map